ENA413166622 Enamine

1H-pyrazole-4,5-diamine

  • MDL number MFCD09473558
  • Cas Number : 16461-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103...105
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142122

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142122
Availability details for
Quantity
  • Availability
  • Add to Cart