ENA413166623 Enamine

{[4-(3-bromophenyl)-1,3-thiazol-2-yl]methyl}(2-methoxyethyl)amine

  • MDL number MFCD18033907
  • Cas Number : 1281656-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142123
Availability details for
Quantity
  • Availability
  • Add to Cart