ENA413166626 Enamine

4-{[6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3-yl]methyl}aniline

  • MDL number MFCD13467475
  • Cas Number : 1094315-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142126
Availability details for
Quantity
  • Availability
  • Add to Cart