ENA413166635 Enamine

methyl 2-(tert-butoxy)propanoate

  • MDL number MFCD16088386
  • Cas Number : 87100-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142142

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142142
Availability details for
Quantity
  • Availability
  • Add to Cart