ENA413166647 Enamine

3-methanesulfonylbenzene-1-sulfonamide

  • MDL number MFCD11205186
  • Cas Number : 22821-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213...215
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142160

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142160
Availability details for
Quantity
  • Availability
  • Add to Cart