ENA413166652 Enamine

1-(3-bromophenyl)-3-propoxypropan-2-amine

  • MDL number MFCD16114941
  • Cas Number : 1248774-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142180

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142180
Availability details for
Quantity
  • Availability
  • Add to Cart