ENA413166655 Enamine

3-hydroxy-4-methanesulfonyl-1$l^{6}-thiolane-1,1-dione

  • MDL number MFCD00195956
  • Cas Number : 53287-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187...189
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142187

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142187
Availability details for
Quantity
  • Availability
  • Add to Cart