ENA413166656 Enamine

N-(2,2-dimethoxyethyl)-3,4-dihydro-2H-pyrrol-5-amine

  • MDL number MFCD15203770
  • Cas Number : 1211463-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139825
Availability details for
Quantity
  • Availability
  • Add to Cart