ENA413166666 Enamine

2,8-dioxaspiro[4.5]decane-1,3-dione

  • MDL number MFCD12088028
  • Cas Number : 5270-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-142216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142216
Availability details for
Quantity
  • Availability
  • Add to Cart