ENA413166668 Enamine

2-(4-bromophenyl)-2-methylpropane-1,3-diol

  • MDL number MFCD21927427
  • Cas Number : 66810-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-142224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142224
Availability details for
Quantity
  • Availability
  • Add to Cart