ENA413166673 Enamine

5-methoxy-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD13175000
  • Cas Number : 139297-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193-194
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142240
Availability details for
Quantity
  • Availability
  • Add to Cart