ENA413166676 Enamine

(1,5-dimethyl-1H-1,3-benzodiazol-2-yl)methanamine

  • MDL number MFCD09882444
  • Cas Number : 887571-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140436

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140436
Availability details for
Quantity
  • Availability
  • Add to Cart