ENA413166677 Enamine

3-fluoro-N-[(3-methyl-1H-pyrazol-4-yl)methyl]pyridin-2-amine

  • MDL number MFCD16859477
  • Cas Number : 1340207-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-142246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142246
Availability details for
Quantity
  • Availability
  • Add to Cart