ENA413166680 Enamine

ethyl 2-[4-(4-methylphenyl)-1,3-thiazol-2-yl]acetate

  • MDL number MFCD09501574
  • Cas Number : 897838-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142266
Availability details for
Quantity
  • Availability
  • Add to Cart