ENA413166681 Enamine

(1-phenylcyclopropyl)methanol

  • MDL number MFCD00001310
  • Cas Number : 31729-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-142267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142267
Availability details for
Quantity
  • Availability
  • Add to Cart