ENA413166686 Enamine

4,4-difluorocyclohexane-1-carbonyl chloride

  • MDL number MFCD13173983
  • Cas Number : 376348-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142276

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-142276
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart