ENA413166688 Enamine

1-bromo-4-cyclopropoxybenzene

  • MDL number MFCD08461662
  • Cas Number : 38380-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142278
Availability details for
Quantity
  • Availability
  • Add to Cart