ENA413166690 Enamine

5-methyl-4-(naphthalen-1-yl)-1,3-thiazol-2-amine

  • MDL number MFCD20171325
  • Cas Number : 107411-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142309
Availability details for
Quantity
  • Availability
  • Add to Cart