ENA413166692 Enamine

(6-bromo-1-methyl-1H-1,3-benzodiazol-2-yl)methanol

  • MDL number MFCD27500721
  • Cas Number : 958863-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142314

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142314
Availability details for
Quantity
  • Availability
  • Add to Cart