ENA413166696 Enamine

3-tert-butyl-6-chloro-[1,2,4]triazolo[4,3-b]pyridazine

  • MDL number MFCD11103591
  • Cas Number : 263401-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151...153
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142318

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142318
Availability details for
Quantity
  • Availability
  • Add to Cart