ENA413166697 Enamine

5-(bromomethyl)-2,2-dimethyloxolane

  • MDL number MFCD20622387
  • Cas Number : 123314-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142319

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142319
Availability details for
Quantity
  • Availability
  • Add to Cart