ENA413166698 Enamine

1-[3-(2-fluorophenyl)-1H-pyrazol-5-yl]piperazine

  • MDL number MFCD17160132
  • Cas Number : 1542693-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142323
Availability details for
Quantity
  • Availability
  • Add to Cart