ENA413166705 Enamine

N-(1-amino-3,3-dimethylbutan-2-yl)-3-methoxyaniline

  • MDL number MFCD21227180
  • Cas Number : 1489008-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60...62
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142336

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142336
Availability details for
Quantity
  • Availability
  • Add to Cart