ENA413166712 Enamine

[2-(cyclopropylamino)ethyl]dimethylamine dihydrochloride

  • MDL number MFCD26936265
  • Cas Number : 1607257-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185...187
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142354
Availability details for
Quantity
  • Availability
  • Add to Cart