ENA413166716 Enamine

methyl 2-(7-amino-2,3-dihydro-1,4-benzodioxin-6-yl)acetate hydrochloride

  • MDL number MFCD26936269
  • Cas Number : 1797842-87-5
  • Molecular Weight : 259.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-(7-amino-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acetate hydrochloride
Inchi Code 1S/C11H13NO4.ClH/c1-14-11(13)5-7-4-9-10(6-8(7)12)16-3-2-15-9;/h4,6H,2-3,5,12H2,1H3;1H
Inchi Key HJWUCRWLWCXKIA-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-142358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142358
Availability details for
Quantity
  • Availability
  • Add to Cart