ENA413166717 Enamine

5-ethyl-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD20654030
  • Cas Number : 144152-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140447
Availability details for
Quantity
  • Availability
  • Add to Cart