ENA413166719 Enamine

1,2-di-tert-butyl pyrrolidine-1,2-dicarboxylate

  • MDL number MFCD24390229
  • Cas Number : 347189-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142397
Availability details for
Quantity
  • Availability
  • Add to Cart