ENA413166722 Enamine

5-(benzyloxy)-2-methanesulfonylpyrimidine

  • MDL number MFCD04109501
  • Cas Number : 16290-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140453

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140453
Availability details for
Quantity
  • Availability
  • Add to Cart