ENA413166732 Enamine

2,5-dichloro-8-fluoroquinoline

  • MDL number MFCD19678387
  • Cas Number : 1342357-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142417

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142417
Availability details for
Quantity
  • Availability
  • Add to Cart