ENA413166738 Enamine

2-(benzylsulfanyl)acetaldehyde

  • MDL number MFCD16096691
  • Cas Number : 7401-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142424

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142424
Availability details for
Quantity
  • Availability
  • Add to Cart