ENA413166740 Enamine

1-(4-ethenylphenyl)ethan-1-one

  • MDL number MFCD09033342
  • Cas Number : 10537-63-0
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-142426

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142426
Availability details for
Quantity
  • Availability
  • Add to Cart