ENA413166742 Enamine

5-[2-(trifluoromethoxy)phenyl]furan-2-carboxylic acid

  • MDL number MFCD02215971
  • Cas Number : 424817-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140458
Availability details for
Quantity
  • Availability
  • Add to Cart