ENA413166746 Enamine

2-[4-(aminomethyl)phenyl]propan-2-ol

  • MDL number MFCD11040533
  • Cas Number : 214758-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142434

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142434
Availability details for
Quantity
  • Availability
  • Add to Cart