ENA413166751 Enamine

1-(2,2,2-trifluoroethyl)azetidine-3-carboxylic acid

  • MDL number MFCD20282477
  • Cas Number : 1339115-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142439
Availability details for
Quantity
  • Availability
  • Add to Cart