ENA413166754 Enamine

2-(methoxymethyl)-1H-imidazole

  • MDL number MFCD00082200
  • Cas Number : 102644-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142444

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142444
Availability details for
Quantity
  • Availability
  • Add to Cart