ENA413166756 Enamine

1-[(piperidin-3-yl)methyl]-1H-1,3-benzodiazole dihydrochloride

  • MDL number MFCD22587612
  • Cas Number : 1351654-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142447
Availability details for
Quantity
  • Availability
  • Add to Cart