ENA413166765 Enamine

4-cyclopropyl-3-methylaniline

  • MDL number MFCD14585533
  • Cas Number : 1208091-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142460

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142460
Availability details for
Quantity
  • Availability
  • Add to Cart