ENA413166767 Enamine

1-(3-methylphenyl)-3-(prop-2-yn-1-yl)urea

  • MDL number MFCD08872860
  • Cas Number : 69921-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142463

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142463
Availability details for
Quantity
  • Availability
  • Add to Cart