ENA413166768 Enamine

2H,3H,5H,6H,7H,8H-pyrido[4,3-c]pyridazin-3-one hydrochloride

  • MDL number MFCD22570206
  • Cas Number : 39716-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142470

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142470
Availability details for
Quantity
  • Availability
  • Add to Cart