ENA413166770 Enamine

3-hydroxy-5-(methoxycarbonyl)benzoic acid

  • MDL number MFCD20645874
  • Cas Number : 167630-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142477
Availability details for
Quantity
  • Availability
  • Add to Cart