ENA413166785 Enamine

2-pentyl-9,10-dihydroanthracene-9,10-dione

  • MDL number MFCD01940478
  • Cas Number : 13936-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142522
Availability details for
Quantity
  • Availability
  • Add to Cart