ENA413166787 Enamine

5-bromo-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD09948014
  • Cas Number : 1016842-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142525

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142525
Availability details for
Quantity
  • Availability
  • Add to Cart