ENA413166791 Enamine

3-chloro-2-(ethanesulfonyl)benzaldehyde

  • MDL number MFCD16619554
  • Cas Number : 1379324-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142530
Availability details for
Quantity
  • Availability
  • Add to Cart