ENA413166792 Enamine

2-(2,2-difluoroethoxy)ethan-1-ol

  • MDL number MFCD19442036
  • Cas Number : 853005-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142533

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-142533
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart