ENA413166793 Enamine

2-(2-chloro-4-fluorophenyl)morpholine

  • MDL number MFCD20296947
  • Cas Number : 1338981-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142539

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142539
Availability details for
Quantity
  • Availability
  • Add to Cart