ENA413166794 Enamine

7-fluoro-5-nitro-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD11897057
  • Cas Number : 1176058-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142540

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142540
Availability details for
Quantity
  • Availability
  • Add to Cart