ENA413166797 Enamine

(4-fluorophenyl)(1-methyl-1H-pyrazol-4-yl)methanamine

  • MDL number MFCD10689688
  • Cas Number : 1152548-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142544

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142544
Availability details for
Quantity
  • Availability
  • Add to Cart