ENA413166799 Enamine

methyl 2-amino-2-(3,4-dichlorophenyl)propanoate

  • MDL number MFCD12151685
  • Cas Number : 1178456-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142546

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142546
Availability details for
Quantity
  • Availability
  • Add to Cart