ENA413166804 Enamine

3-methyl-5-phenyl-1H-pyrazol-4-amine dihydrochloride

  • MDL number MFCD02067734
  • Cas Number : 305382-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142577
Availability details for
Quantity
  • Availability
  • Add to Cart