ENA413166806 Enamine

4-fluoro-7-nitro-2,3-dihydro-1H-indole

  • MDL number MFCD12405174
  • Cas Number : 1167056-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142580
Availability details for
Quantity
  • Availability
  • Add to Cart